3,3-Dimethyl-2-oxobutyric acid ∼60% in aqueous solution
Supplier: Thermo Scientific
Danger
Synonyms: 3,3-Dimethyl-2-oxobutanoic acid, Trimethylpyruvic Acid
B25582.18 B25582.30
B25582.18EA 40.4 EUR
B25582.18 B25582.30
3,3-Dimethyl-2-oxobutyric acid ∼60% in aqueous solution
3,3-Dimethyl-2-oxobutyric acid
CAS No.: 815-17-8 Molecular Formula: C6H10O3 Formula Weight: 130.14 UN No.: UN3265 MDL No.: MFCD00154352
Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00154352
CAS Number: 815-17-8
EINECS: 212-418-2
UN: 3265
ADR: 8,III