Ethyl-3-aminobenzofuran-2-carboxylate 97%

Supplier: Thermo Scientific

Synonyms: Ethyl 3-Aminocoumarilate, 3-Aminobenzofuran-2-carboxylic acid Ethyl Ester, 3-Aminocoumarilic acid Ethyl Ester, Ethyl 3-aminobenzofuran-2-carboxylate

H27124.06 H27124.03
H27124.06EA 107 EUR
H27124.06 H27124.03
Ethyl-3-aminobenzofuran-2-carboxylate 97%
Ethyl-3-aminobenzofuran-2-carboxylate

Purity: 97% CAS No.: 39786-35-1 Molecular Formula: C11H11NO3 Formula Weight: 205.22 Melting Point: 77-82° MDL No.: MFCD00641330

Formula: C₁₁H₁₁NO₃
MW: 205.21 g/mol
Melting Pt: 77…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00641330
CAS Number: 39786-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR