1-Amino-1-cyclopropanecarbonitrile hydrochloride 97%
Supplier: Thermo Fisher Scientific
Danger
H27288.03 H27288.06
H27288.03EA 41.6 EUR
H27288.03 H27288.06
1-Amino-1-cyclopropanecarbonitrile hydrochloride 97%
1-Amino-1-cyclopropanecarbonitrile hydrochloride
Purity: 97% CAS No.: 127946-77-4 Molecular Formula: C4H6N2.HCl Formula Weight: 118.56 Melting Point: ca 220° dec. UN No.: UN3439 MDL No.: MFCD04114063
Formula: C₄H₇ClN₂
MW: 118.56 g/mol
Melting Pt: ∼ 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD04114063
CAS Number: 127946-77-4
ADR: 6.1 III