4-Benzyloxy-2-chloropyrimidine 95%
Supplier: Thermo Scientific
|
Warning
|
H27313.03EA
301
EUR
H27313.03
H27313.MD
4-Benzyloxy-2-chloropyrimidine 95%
4-Benzyloxy-2-chloropyrimidine
Formula:
C₁₁H₉ClN₂O MW: 220,66 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06798233 CAS Number: 108381-28-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...