4-(4-Dimethylaminophenylazo)benzoic acid sodium salt indicator
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
p-Methyl red, Sodium 4-(4-dimethylaminophenylazo)benzoate
H27830.06
H27830.06EA
209
EUR
H27830.06
4-(4-Dimethylaminophenylazo)benzoic acid sodium salt indicator
4-(4-Dimethylaminophenylazo)benzoic acidsodium salt
CAS No.: 845-46-5
Molecular Formula: C15H14N3NaO2
Formula Weight: 291.28
Melting Point:
Formula:
C₁₅H₁₄N₃O₂Na MW: 291.28 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020350 CAS Number: 845-46-5 |