Chloro(p-cymene)-N-(p-toluenesulphonyl)(R,R)-1,2-cyclohexanediamineruthenium(I) 97%
Supplier: Thermo Fisher Scientific
Warning
H30264.MD H30264.03
H30264.MDEA 123 EUR
H30264.MD H30264.03
Chloro(p-cymene)-N-(p-toluenesulphonyl)(R,R)-1,2-cyclohexanediamineruthenium(I) 97%
Chloro(p-cymene)-N-(p-toluenesulphonyl)(R,R)-1,2-cyclohexanediamineruthenium(I)
Purity: 97% CAS No.: 213603-12-4 Molecular Formula: C23H33ClN2O2RuS Formula Weight: 538.11 MDL No.: MFCD12546183
Formula: C₂₃H₃₃ClN₂O₂RuS
MW: 538.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546183
CAS Number: 213603-12-4