Ethyl-2-cyano-2-methylpropionate 97%
Supplier: Thermo Scientific
Danger
Synonyms: Ethyl 2-Cyano-2-methylpropionate, 2-Cyano-2-methylpropionic Acid Ethyl Ester
H31432.09 H31432.18
H31432.09EA 47.1 EUR
H31432.09 H31432.18
Ethyl-2-cyano-2-methylpropionate 97%
Ethyl-2-cyano-2-methylpropionate
Purity: 97% CAS No.: 1572-98-1 Molecular Formula: C7H11NO2 Formula Weight: 141.17 Boiling Point: 185° MDL No.: MFCD00034667
Formula: C₇H₁₁NO₂
MW: 141.17 g/mol
Boiling Pt: 185 °C
Density: 0.97
Storage Temperature: Ambient
MDL Number: MFCD00034667
CAS Number: 1572-98-1