1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1,2,3,4-Tetrahydro-1-naphthoic acid
H32138.MD H32138.03
H32138.MDEA 91.4 EUR
H32138.MD H32138.03
1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid 98%
1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid
Purity: 98% CAS No.: 1914-65-4 Molecular Formula: C11H12O2 Formula Weight: 176.22 Melting Point: 82-85° MDL No.: MFCD02179127
Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Melting Pt: 82…85 °C
Storage Temperature: Ambient
MDL Number: MFCD02179127
CAS Number: 1914-65-4