Erbium ≥99.9% (REO, rare earth oxide basis), powder -40 mesh

Supplier: Thermo Scientific
Danger


Packaged under argon.

44169.06EA 272 EUR
44169.06 44169.14
Erbium ≥99.9% (REO, rare earth oxide basis), powder -40 mesh
Erbium
Formula: Er
MW: 167,26 g/mol
Boiling Pt: 2863 °C (1013 hPa)
Melting Pt: 1529 °C
Density: 9,066 g/cm³ (20 °C)
Storage Temperature: Ambient
UN: 3089
ADR: 4.1,II
Merck Index: 12,03679

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR