4,5-Dimethoxy-2-nitrophenylacetonitrile 98%
Supplier: Thermo Scientific
Danger
Synonyms: 2-(4,5-Dimethoxy-2-nitrophenyl)acetonitrile
H32747.MD H32747.03
H32747.MDEA 42.4 EUR
H32747.MD H32747.03
4,5-Dimethoxy-2-nitrophenylacetonitrile 98%
4,5-Dimethoxy-2-nitrophenylacetonitrile
Purity: 98% CAS No.: 17354-04-0 Molecular Formula: C10H10N2O4 Formula Weight: 222.20 Melting Point: 110-112° UN No.: UN3439 MDL No.: MFCD00033967
Formula: C₁₀H₁₀N₂O₄
MW: 222.20 g/mol
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00033967
CAS Number: 17354-04-0
ADR: 6.1,III