2-Fluoro-4-(trifluoromethoxy)benzaldehyde 97%
Supplier: Thermo Fisher Scientific
Warning
H33203.03 H33203.06
H33203.03EA 96.9 EUR
H33203.03 H33203.06
2-Fluoro-4-(trifluoromethoxy)benzaldehyde 97%
2-Fluoro-4-(trifluoromethoxy)benzaldehyde
Purity: 97% CAS No.: 1227628-83-2 Molecular Formula: C8H4F4O2 Formula Weight: 208.11 Refractive Index: 1.4455 MDL No.: MFCD16652440
Formula: C₈H₄F₄O₂
MW: 208.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652440
CAS Number: 1227628-83-2