Ethyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate 96%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Ethyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate
H34116.03
H34116.06
H34116.03EA
44.1
EUR
H34116.03
H34116.06
Ethyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate 96%
Ethyl-2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate
Purity: 96%
CAS No.: 40106-13-6
Molecular Formula: C12H17NO2S
Formula Weight: 239.33
Melting Point: 87-88°
MDL No.: MFCD00216932
Formula:
C₁₂H₁₇NO₂S Melting Pt: 87…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216932 CAS Number: 40106-13-6 |