Palladium 10% on activated carbon paste (50% water) Type 39
Supplier: Thermo Fisher Scientific
H37431.06 H37431.14 H37431.22
H37431.06EA 185 EUR
H37431.06 H37431.14 H37431.22
Palladium 10% on activated carbon paste (50% water) Type 39
Palladium
CAS No.: 7440-05-3 Physical Form: Solid Appearance: Dark grey Odour: Odourless MDL No.: MFCD03457879
Formula: Pd
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
ADR: 4.1,III