Ethyl-3-tert-butyl-1H-pyrazole-5-carboxylate 97%
Supplier: Thermo Scientific
Warning
Synonyms: 3-tert-Butyl-1H-pyrazole-5-carboxylic acid ethyl ester
H50678.03 H50678.06
H50678.03EA 96.8 EUR
H50678.03 H50678.06
Ethyl-3-tert-butyl-1H-pyrazole-5-carboxylate 97%
Ethyl-3-tert-butyl-1H-pyrazole-5-carboxylate
Purity: 97% CAS No.: 916791-97-4 Molecular Formula: C10H16N2O2 Formula Weight: 196.25 MDL No.: MFCD00173782
Formula: C₁₀H₁₆N₂O₂
MW: 196.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173782
CAS Number: 916791-97-4