2,5-Bis(3,7-dimethyloctyloxy)terephthalaldehyde 98%
Supplier: Thermo Scientific
Warning
H51067.MD H51067.03
H51067.MDEA 49 EUR
H51067.MD H51067.03
2,5-Bis(3,7-dimethyloctyloxy)terephthalaldehyde 98%
2,5-Bis(3,7-dimethyloctyloxy)terephthalaldehyde
Purity: 98% CAS No.: 325461-35-6 Molecular Formula: C28H46O4 Formula Weight: 446.66 MDL No.: MFCD03427237
Formula: C₂₈H₄₆O₄
MW: 446.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03427237
CAS Number: 325461-35-6