(1S,3R)-3-Aminocyclopentanecarboxylic acid 95%
Supplier: Thermo Fisher Scientific
Warning
H52040.MD H52040.03
H52040.MDEA 123 EUR
H52040.MD H52040.03
(1S,3R)-3-Aminocyclopentanecarboxylic acid 95%
(1S,3R)-3-Aminocyclopentanecarboxylic acid
Purity: 95% CAS No.: 71830-07-4 Molecular Formula: C6H11NO2 Formula Weight: 129.16 MDL No.: MFCD00211290
Formula: C₆H₁₁NO₂
MW: 129.159 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00211290
CAS Number: 71830-07-4