tert-Butyl-3-(aminomethyl)piperidine-1-carboxylate 97%
Supplier: Thermo Scientific
Warning
Synonyms: tert-Butyl 3-(aminomethyl)piperidine-1-carboxylate, tert-Butyl 3-(Aminomethyl)-1-piperidinecarboxylate, 3-(Aminomethyl)-1-Boc-piperidine, 3-(Aminomethyl)-1-tert-butoxycarbonylpiperidine, 3-(Aminomethyl)-1-piperidinecarboxylic acid tert-Butyl ester
H52832.03 H52832.06
H52832.03EA 62.9 EUR
H52832.03 H52832.06
tert-Butyl-3-(aminomethyl)piperidine-1-carboxylate 97%
tert-Butyl-3-(aminomethyl)piperidine-1-carboxylate
Purity: 97% CAS No.: 162167-97-7 Molecular Formula: C11H22N2O2 Formula Weight: 214.31 MDL No.: MFCD01317792
Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01317792
CAS Number: 162167-97-7