Silver ≥99.9% (metals basis), wire 0.404 mm (0.0159 in) Ø, annealed
Supplier: Thermo Fisher Scientific
45852.G9 45852.H4
45852.G9EA 363 EUR
45852.G9 45852.H4
Silver ≥99.9% (metals basis), wire 0.404 mm (0.0159 in) Ø, annealed
Silver
Purity: 99.9% CAS No.: 7440-22-4 Physical Form: ≈1.35g/m Odour: Characteristic MDL No.: MFCD00003397
Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 962 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647