1,3-Diphenyl-2-propyn-1-ol 90%, tech.
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1,3-diphenylprop-2-yn-1-ol
H53374.03 H53374.06
H53374.03EA 38.5 EUR
H53374.03 H53374.06
1,3-Diphenyl-2-propyn-1-ol 90%, tech.
1,3-Diphenyl-2-propyn-1-ol
Purity: 90% CAS No.: 1817-49-8 Molecular Formula: C15H12O Formula Weight: 208.26 Flash Point: 110°(230°F) Refractive Index: 1.618 UN No.: UN3082 MDL No.: MFCD06654198
Formula: C₁₅H₁₂O
MW: 208.26 g/mol
Density: 1.099
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD06654198
CAS Number: 1817-49-8
UN: 3082
ADR: 9 III