1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol 97%
Supplier: Thermo Fisher Scientific
Warning
H53435.06 H53435.14
H53435.06EA 88.2 EUR
H53435.06 H53435.14
1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol 97%
1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol
Purity: 97% CAS No.: 214746-69-7 Molecular Formula: C20H21NO Formula Weight: 291.39 Melting Point: 142-144° MDL No.: MFCD17677347
Formula: C₂₀H₂₁NO
MW: 291.39 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD17677347
CAS Number: 214746-69-7