cis-5-Norbornene exo-2,3-dicarboxylic anhydride 95%
Supplier: Thermo Scientific
Danger
Synonyms: (3aR,4R,7S,7aS)-rel-3a,4,7,7a-Tetrahydro-4,7-methanoisobenzofuran-1,3-dione, Bicyclo[2.2.1]hept-5-ene-exo-2,3-dicarboxylic Anhydride
H53446.06 H53446.14
H53446.06EA 202 EUR
H53446.06 H53446.14
cis-5-Norbornene exo-2,3-dicarboxylic anhydride 95%
cis-5-Norbornene exo-2,3-dicarboxylic anhydride
Purity: 95% CAS No.: 2746-19-2 Molecular Formula: C9H8O3 Formula Weight: 164.16 Melting Point: 140-145° MDL No.: MFCD00630580
Formula: C₉H₈O₃
MW: 164.16 g/mol
Melting Pt: 140…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00630580
CAS Number: 2746-19-2
EINECS: 220-384-5