1,1-Dimethylpropargylamine 95%
Supplier: Thermo Scientific
Danger
Synonyms: 2-Methylbut-3-yn-2-amine
H53448.06 H53448.14
H53448.06EA 74.4 EUR
H53448.06 H53448.14
1,1-Dimethylpropargylamine 95%
1,1-Dimethylpropargylamine
Purity: 95% CAS No.: 2978-58-7 Molecular Formula: C5H9N Formula Weight: 83.13 Flash Point: 2°(36°F) Refractive Index: 1.4237 Boiling Point: 79-80° UN No.: UN2733 MDL No.: MFCD00008052
Formula: C₅H₉N
MW: 83.134 g/mol
Boiling Pt: 79…80 °C
Density: 0.79
Flash Pt: 2 °C (36 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008052
CAS Number: 2978-58-7
EINECS: 221-029-7
ADR: 3,II