Ethyl-2-methylpyridine-4-carboxylate hydrochloride 95%
Supplier: Thermo Scientific
Danger
H57239.03 H57239.06
H57239.03EA 62.5 EUR
H57239.03 H57239.06
Ethyl-2-methylpyridine-4-carboxylate hydrochloride 95%
Ethyl-2-methylpyridine-4-carboxylate hydrochloride
Purity: 95% CAS No.: 65550-32-5 Molecular Formula: C9H11NO2.HCl Formula Weight: 201.65 Melting Point: 145-151° MDL No.: MFCD09991690
Formula: C₉H₁₁NO₂·HCl
MW: 201.65 g/mol
Melting Pt: 145…151 °C
Storage Temperature: Ambient
MDL Number: MFCD09991690
CAS Number: 65550-32-5