p-Toluenesulphonic acid monohydrate 98
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 4-Methylbenzenesulphonic acid hydrate, Toluene-4-sulphonic acid monohydrate
A14119.0B A14119.0I A14119.30
A14119.0BEA 66.8 EUR
A14119.0B A14119.0I A14119.30
p-Toluenesulphonic acid monohydrate 98
p-Toluenesulphonic acid monohydrate
Formula: C₇H₈O₃S·1H₂O
MW: 190.22 g/mol
Boiling Pt: 140 °C (20 mmHg)
Melting Pt: 103…106 °C (anhydr.)
Density: 1.24
Flash Pt: 150 °C (302 °F)
Storage Temperature: Ambient
MDL Number: MFCD00142137
CAS Number: 6192-52-5
EINECS: 203-180-0
UN: 2585
ADR: 8 III
Merck Index: 12,09671