tert-Butyl (S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride 98%
Supplier: Thermo Fisher Scientific
H64468.03
H64468.06
H64468.03EA
85.7
EUR
H64468.03
H64468.06
tert-Butyl (S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride 98%
tert-Butyl (S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride
Purity: 99%
CAS No.: 77497-74-6
Molecular Formula: C14H19NO2.HCl
Formula Weight: 269.77
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 5744730
MDL No.: MFCD04114841
Formula:
C₁₄H₁₉NO₂ MW: 233.311 g/mol |
MDL Number:
MFCD04114841 CAS Number: 77497-74-6 |