(2-Hydroxy-3-N,N,N-trimethylamino)propyl β-cyclodextrin chloride hydrate
Supplier: Thermo Scientific
Synonyms: (2-Hydroxy-3-N,N,N-trimethylamino)propyl beta-cyclodextrin chloride hydrate, Quaternary ammonium beta-cylodextrin, QA-beta-CD
H27604.MD H27604.03
H27604.MDEA 84.4 EUR
H27604.MD H27604.03
(2-Hydroxy-3-N,N,N-trimethylamino)propyl β-cyclodextrin chloride hydrate
(2-Hydroxy-3-N,N,N-trimethylamino)propyl β-cyclodextrin chloride hydrate
Melting Point: ca 280° dec. MDL No.: MFCD00078139
Melting Pt: ∼ 280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00078139
EINECS: 231-493-2