N,N-Dimethylpropionamide 98+%
Supplier: Thermo Fisher Scientific
|
Warning
|
Purity: 98+%
CAS No.: 758-96-3
Molecular Formula: C5H11NO
Formula Weight: 101.15
Flash Point: 62°(144°F)
Physical Form: Liquid
Density: 0.920
Refractive Index: 1.4400
Appearance: Colourless
Melting Point: -45° lit.
Boiling Point: 174-175° lit.
Beilstein Reference: 1740719
MDL No.: MFCD00009301
Formula:
C₅H₁₁NO Boiling Pt: 174…175 °C (lit.) Melting Pt: –45 °C (lit.) Density: 0.920 Flash Pt: 62 °C (144 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009301 CAS Number: 758-96-3 EINECS: 212-064-9 |