D-Amygdalin 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Laetrile, Amygdalin, Amygdaloside, D-Mandelonitrile 6-O-β-D-glucosido-β-D-glucoside, (R)-2-Phenyl-2-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)acetonitrile, D-Mandelonitrile-β-gentiobioside
Purity: 98%
CAS No.: 29883-15-6
Molecular Formula: C20H27NO11
Formula Weight: 457.44
Physical Form: Powder
Appearance: White to off-white
Melting Point: 210-215°
Beilstein Reference: 66856
Optical Rotation: -38° (c=1.2 in water)
MDL No.: MFCD00006598
Applications: A benzylic glycoside that has been used as an antineoplastic agent
Formula:
C₂₀H₂₇NO₁₁ MW: 457.43 g/mol Melting Pt: 210…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006598 CAS Number: 29883-15-6 EINECS: 249-925-3 Merck Index: 14,597 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...