Phosphoenol pyruvic acid trisodium salt hydrate 98%
Supplier: Thermo Fisher Scientific
J63013.03 J63013.06
J63013.03EA 180 EUR
J63013.03 J63013.06
Phosphoenol pyruvic acid trisodium salt hydrate 98%
Phosphoenol pyruvic acid trisodium salt hydrate
Purity: 98% CAS No.: 5541-93-5 Molecular Formula: C3H2Na3O6P.7H2O Formula Weight: 360.09 Physical Form: Powder Appearance: White MDL No.: MFCD00150737
Applications: When coupled with pyruvate kinase it completes an ATP-regenerating system
Formula: C₃H₂Na₃O₆P
MW: 233.986 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150737
CAS Number: 5541-93-5
EINECS: 226-906-8