Nabumetone
Supplier: Thermo Fisher Scientific
Warning
|
Synonyms:
4-(6-Methoxynaphthalen-2-yl)butan-2-one
J63072.06
J63072.14
J63072.06EA
104
EUR
J63072.06
J63072.14
Nabumetone
Nabumetone
CAS No.: 42924-53-8
Molecular Formula: C15H16O2
Formula Weight: 228.29
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Appearance: White
Melting Point: 76-78°
Solubility: Soluble in alcohol or chloroform
MDL No.: MFCD00079518
Applications: Anti-inflammatory, anti-bacterial. A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase
Formula:
C₁₅H₁₆O₂ Melting Pt: 76…78 °C |
MDL Number:
MFCD00079518 CAS Number: 42924-53-8 Merck Index: 14,6343 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...