(+)-Dehydroabietylamine hydrochloride
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Leelamine hydrochloride, Dehydroabietylamine hydrochloride, 1R,2,3,4,4aS,9,10,10aR-octahydro-1,4a-dimethyl-7-(1-methylethyl)-1-phenanthrenemethanamine, hydrochloride
J63123.MA
J63123.MF
J63123.MAEA
136
EUR
J63123.MA
J63123.MF
(+)-Dehydroabietylamine hydrochloride
(+)-Dehydroabietylamine hydrochloride
CAS No.: 16496-99-4
Molecular Formula: C20H31N.HCl
Formula Weight: 321.90
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White
Beilstein Reference: 3084620
MDL No.: MFCD00213430
Applications: Resolving agent for carboxylic acids
Formula:
C₂₀H₃₁N·HCl |
MDL Number:
MFCD00213430 CAS Number: 16496-99-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...