N,N,N',N'-Tetrakis(2-pyridylmethyl)ethylenediamine

Supplier: Thermo Scientific
Warning

J64206.MCEA 142 EUR
J64206.MC
N,N,N',N'-Tetrakis(2-pyridylmethyl)ethylenediamine
N,N,N',N'-Tetrakis(2-pyridylmethyl)ethylenediamine

Applications: A cell permeable and extremely high-affinity chelator of heavy metals

Formula: C₂₆H₂₈N₆
MW: 424,55 g/mol
Melting Pt: 110…112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036918
CAS Number: 16858-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR