Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 10 nm OD1 (Abs. 520 nm)
Supplier: Thermo Fisher Scientific
J67188.AC J67188.AE
J67188.ACEA 92.7 EUR
J67188.AC J67188.AE
Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 10 nm OD1 (Abs. 520 nm)
Gold
CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Pink to red Odour: Odourless MDL No.: MFCD00003436
Applications: For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Refrigerator
MDL Number: MFCD00003436
CAS Number: 7440-57-5
Merck Index: 12,04526