Chloroform for molecular biology
Supplier: Thermo Fisher Scientific
|
Danger
|
CAS No.: 67-66-3
Molecular Formula: CHCl3
Formula Weight: 119.38
Physical Form: Liquid
Density: 1.492
Refractive Index: 1.446
Appearance: Colourless
Odour: Characteristic
Melting Point: -63°
Boiling Point: 61°
Beilstein Reference: 1731042
UN No.: UN1888
MDL No.: MFCD00000826
Formula:
CHCl₃ Boiling Pt: 61 °C Melting Pt: –63 °C Density: 1.492 Storage Temperature: Ambient |
MDL Number:
MFCD00000826 CAS Number: 67-66-3 EINECS: 200-663-8 UN: 1888 ADR: 6.1,III Merck Index: 14,2141 |