3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) for electrophoresis
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulphonate, CHAPS
J67359.06
J67359.09
J67359.14
J67359.06EA
185
EUR
J67359.06
J67359.09
J67359.14
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) for electrophoresis
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
CAS No.: 75621-03-3
Molecular Formula: C32H58N2O7S
Formula Weight: 614.88
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 156-158°
MDL No.: MFCD00012116
Formula:
C₃₂H₅₈N₂O₇S Melting Pt: 156…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081083 CAS Number: 75621-03-3 |