2,2'-Dinaphthyl ether ≥98%
Supplier: Thermo Fisher Scientific
L02565.06 L02565.14
L02565.06EA 56.5 EUR
L02565.06 L02565.14
2,2'-Dinaphthyl ether ≥98%
2,2'-Dinaphthyl ether
Purity: 98+% CAS No.: 613-80-9 Molecular Formula: C20H14O Formula Weight: 270.33 Melting Point: 103-105° Beilstein Reference: 2054097 MDL No.: MFCD00015447
Formula: C₂₀H₁₄O
MW: 270.33 g/mol
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00015447
CAS Number: 613-80-9
EINECS: 210-356-0