N-Methyltrimethylacetamide 96%

Supplier: Thermo Fisher Scientific

L08916.06 L08916.14
L08916.06EA 78.1 EUR
L08916.06 L08916.14
N-Methyltrimethylacetamide 96%
N-Methyltrimethylacetamide

Purity: 96% CAS No.: 6830-83-7 Molecular Formula: C6H13NO Formula Weight: 115.18 Melting Point: 89-93° Boiling Point: 105-110°/15mm Beilstein Reference: 1744142 MDL No.: MFCD00043643

Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 105…110 °C (15 mmHg)
Melting Pt: 89…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00043643
CAS Number: 6830-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR