Diethyl ethylidenemalonate 99%

Supplier: Thermo Fisher Scientific

Synonyms: Diethyl 2-ethylidenemalonate

L12117.06 L12117.03
L12117.06EA 69 EUR
L12117.06 L12117.03
Diethyl ethylidenemalonate 99%
Diethyl ethylidenemalonate

Purity: 99% CAS No.: 1462-12-0 Molecular Formula: C9H14O4 Formula Weight: 186.21 Flash Point: 110°(230°F) Refractive Index: 1.4420 Boiling Point: 115-116°/17mm Beilstein Reference: 1773932 MDL No.: MFCD00009145

Formula: C₉H₁₄O₄
MW: 186.21 g/mol
Boiling Pt: 115…116 °C (17 mmHg)
Density: 1.019
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009145
CAS Number: 1462-12-0
EINECS: 215-965-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR