Ethyl L-2-isothiocyanatopropionate 97%

Supplier: Thermo Fisher Scientific
Danger

L12532.03 L12532.06
L12532.03EA 80.8 EUR
L12532.03 L12532.06
Ethyl L-2-isothiocyanatopropionate 97%
Ethyl L-2-isothiocyanatopropionate

Purity: 97% CAS No.: 39574-16-8 Molecular Formula: C6H9NO2S Formula Weight: 159.20 Storage Temperature: 2°C to 8°C Refractive Index: 1.4925 Boiling Point: 63-64°/1mm Beilstein Reference: 1723243 UN No.: UN2922 MDL No.: MFCD00070636

Formula: C₆H₉NO₂S
Boiling Pt: 63…64 °C (1 mmHg)
MDL Number: MFCD09025691
CAS Number: 39574-16-8
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR