1-Isopropenyl-2-benzimidazolidinone ≥98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1-(Prop-1-en-2-yl)-2,3-dihydro-1H-1,3-benzodiazol-2-one, 1-(Prop-1-en-2-yl)-1H-benzo[d]imidazol-2(3H)-one
L17626.03 L17626.06
L17626.03EA 46.1 EUR
L17626.03 L17626.06
1-Isopropenyl-2-benzimidazolidinone ≥98%
1-Isopropenyl-2-benzimidazolidinone
Purity: 98+% CAS No.: 52099-72-6 Molecular Formula: C10H10N2O Formula Weight: 174.20 Melting Point: 121-123° Beilstein Reference: 140587 MDL No.: MFCD00218253
Formula: C₁₀H₁₀N₂O
MW: 174.203 g/mol
Melting Pt: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00218253
CAS Number: 52099-72-6
EINECS: 257-661-5