Morpholinosulphur trifluoride 95%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Morph-DAST
L19751.03
L19751.06
L19751.14
L19751.03EA
43.1
EUR
L19751.03
L19751.06
L19751.14
Morpholinosulphur trifluoride 95%
Morpholinosulphur trifluoride
Purity: 95%
CAS No.: 51010-74-3
Molecular Formula: C4H8F3NOS
Formula Weight: 175.17
Storage Temperature: -30°C to -10°C
Flash Point: 76°(168°F)
Refractive Index: 1.4545
Boiling Point: 41-42°/0.5mm
Beilstein Reference: 606224
UN No.: UN3265
MDL No.: MFCD00037057
Formula:
C₄H₈F₃NOS MW: 175.17 g/mol Boiling Pt: 41…42 °C (0.5 mmHg) Density: 1.436 Flash Pt: 76 °C (168 °F) |
MDL Number:
MFCD00037057 CAS Number: 51010-74-3 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...