Tetramethylammonium hydroxide 25% in aqueous solution (w/w)
Supplier: Thermo Scientific
Danger
L09991.AK L09991.AC L09991.AU
L09991.AKEA 61.9 EUR
L09991.AK L09991.AC L09991.AU
Tetramethylammonium hydroxide 25% in aqueous solution (w/w)
Tetramethylammonium hydroxide
Formula: C₄H₁₃NO
MW: 91.15 g/mol
Boiling Pt: ∼102 °C
Density: 1.014
Storage Temperature: Ambient
MDL Number: MFCD00008280
CAS Number: 75-59-2
EINECS: 200-882-9
UN: 1835
ADR: 8,II
Merck Index: 14,9224