Dodecamolybdophosphoric acid hydrate (>47.5% Mo)
Supplier: Thermo Scientific
Danger
Synonyms: Molybdophosphoric acid hydrate, 12-Molybdophosphoric acid hydrate, Phosphomolybdic acid hydrate
L15827.14 L15827.22 L15827.36
L15827.14EA 37 EUR
L15827.14 L15827.22 L15827.36
Dodecamolybdophosphoric acid hydrate (>47.5% Mo)
Dodecamolybdophosphoric acid hydrate
CAS No.: 51429-74-4 Molecular Formula: H3PO4.12MoO3.xH2O Formula Weight: 1825.25(anhy) Melting Point: 78-90° Merck Reference: 14,7342 UN No.: 3085 MDL No.: MFCD00149913
Formula: H₃[P(Mo₃O₁₀)₄]·nH₂O
MW: 1843.27 g/mol
Melting Pt: 78…90 °C
Density: 3.15
Storage Temperature: Ambient
MDL Number: MFCD00149913
CAS Number: 51429-74-4
EINECS: 234-713-5
UN: 3085
ADR: 5.1,III
Merck Index: 14,7342