1,1,1-Trichloro-2-methyl-2-propanol hemihydrate ≥98.0% NF
Supplier: Spectrum Chemical
Warning
Synonyms: 1,1,1-Trichloro-2-methyl-2-propanol Hemihydrate, Chlorbutanol hemihydrate, Chlorobutanol hemihydrate, Chlorobutanol hydrous
CH122-12KGBL CH122-2.5KG CH122-50KGBL
SPCMCH122-12KGBLEA 0 EUR
SPCMCH122-12KGBL SPCMCH122-2.5KG SPCMCH122-50KGBL
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate ≥98.0% NF
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate
Formula: C₈H₁₆Cl₆O₃
MW: 372.93 g/mol
Boiling Pt: 167 °C (1013 hPa)
Melting Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD02179352
CAS Number: 6001-64-5
EINECS: 200-317-6
Order Now
Specification Test Results
Assay 98% Min.