1,1,1-Trichloro-2-methyl-2-propanol, anhydrous NF
Supplier: Spectrum Chemical
Synonyms: Chlorbutanol, 1,1,1-Trichloro-2-methylpropan-2-ol, Chlorobutanol
CH123-12KGBL CH123-50KGBL CH123-25GM CH123-500GM
SPCMCH123-12KGBLEA 0 EUR
SPCMCH123-12KGBL SPCMCH123-50KGBL SPCMCH123-25GM SPCMCH123-500GM
1,1,1-Trichloro-2-methyl-2-propanol, anhydrous NF
1,1,1-Trichloro-2-methyl-2-propanol
Melamine statement (available upon request)
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formula: C₄H₇Cl₃O
Boiling Pt: 166 °C (1013 hPa)
Melting Pt: 78 °C
MDL Number: MFCD02179352
CAS Number: 57-15-8
EINECS: 200-317-6
Merck Index: 13,02148
Order Now
Specification Test Results
Assay 98% Min.