Pyridine-2,6-dicarboxylic acid
Supplier: Spectrum Chemical
Warning
D2343-1KG D2343-25KGBL D2343-50KGBL
SPCMD2343-1KGEA 1440 EUR
SPCMD2343-1KG SPCMD2343-25KGBL SPCMD2343-50KGBL
Pyridine-2,6-dicarboxylic acid
Pyridine-2,6-dicarboxylic acid
Formula: C₇H₅NO₄
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 327 °C
Flash Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00006299
CAS Number: 499-83-2
EINECS: 207-894-3
Order Now
Specification Test Results
Assay 98% Min.