4-Hydroxy-4-methyl-2-pentanone
Supplier: Spectrum Chemical
Danger
DI102-200LTBL DI102-20LTMT
SPCMDI102-200LTBLEA 6430 EUR
SPCMDI102-200LTBL SPCMDI102-20LTMT
4-Hydroxy-4-methyl-2-pentanone
4-Hydroxy-4-methyl-2-pentanone
Formula: (CH₃)₂C(OH)CH₂COCH₃
Boiling Pt: 166 °C (1013 hPa)
Melting Pt: –44 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00004471
CAS Number: 123-42-2
EINECS: 204-626-7
UN: 1148
ADR: 3,III
REACH: 01-2119473975-21
Merck Index: 13,02983
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2da78bff918d252787ee81ab8857bd3b19e51ac&vid=d8a0cccff2cac15b826ba733a013451ac11e3b51&prid=web&clid=SAPDAM