Hydroquinone USP
Supplier: Spectrum Chemical
Danger
Synonyms: 1,4-Dihydroxybenzene, Quinol
HY113-2.5KG HY113-25KGBL HY113-50KGBL HY113-500GM
SPCMHY113-2.5KGEA 4120 EUR
SPCMHY113-2.5KG SPCMHY113-25KGBL SPCMHY113-50KGBL SPCMHY113-500GM
Hydroquinone USP
Hydroquinone
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formula: C₆H₄(OH)₂
MW: 110.11 g/mol
Boiling Pt: 285
Melting Pt: 172
Density: 1.332 g/cm³ (20 °C)
Flash Pt: 165
Storage Temperature: Ambient
MDL Number: MFCD00002339
CAS Number: 123-31-9
EINECS: 204-617-8
UN: 3077
ADR: 9 III
Merck Index: 13,04833
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=203cfb319003b2b4489cbe3ab15876fc37f27cb8&vid=6d76260c677491e292c03637f8a08e9194282d48&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 99% Min.