Isopropyl palmitate NF
Supplier: Spectrum Chemical
IS160-200LTBL IS160-20LTMT IS160-4LTGL IS160-500MLGL
SPCMIS160-200LTBLEA 0 EUR
SPCMIS160-200LTBL SPCMIS160-20LTMT SPCMIS160-4LTGL SPCMIS160-500MLGL
Isopropyl palmitate NF
Isopropyl palmitate
Formula: C₁₉H₃₈O₂
Boiling Pt: 160 °C (2 mmHg)
Melting Pt: 13.5 °C
Density: 0.855 g/cm³ (20 °C)
MDL Number: MFCD00008993
CAS Number: 142-91-6
EINECS: 205-571-1
Order Now
Specification Test Results
Assay 90% Min.