L(+)-Leucine USP
Supplier: Spectrum Chemical
Synonyms: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine
L1202-1KG L1202-25KGBL L1202-5KG
SPCML1202-1KGEA 516 EUR
SPCML1202-1KG SPCML1202-25KGBL SPCML1202-5KG
L(+)-Leucine USP
L(+)-Leucine
Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Melting Pt: 300
Density: 1.165 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002617
CAS Number: 61-90-5
EINECS: 200-522-0
Merck Index: 14,05451
Order Now
Specification Test Results
Assay 98.5% Min.